Assay Details
Amplicon Length: | 96 |
Chromosome Location: | Chr. 16 - 56858319 - 56858319 [+] on 56858319 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-138-2-3p | MIMAT0004596 |
| Human | hsa-miR-138-5p | MIMAT0000430 |
Stem-loop Details
Human

Genomic Map