Assay Details
Gene Family ID: | MIPF0000075, mir-138 |
Mature miRNA Sequence: | GCUAUUUCACGACACCAGGGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-138-2-3p | MIMAT0004596 |
| Dasypus novemcinctus | dno-miR-138-2-3p | MIMAT0047650 |
| Pteropus alecto | pal-miR-138-2-3p | MIMAT0040153 |
Stem-loop Details
Human
Dasypus novemcinctus
Pteropus alecto

Genomic Map