Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-320a-3p | MIMAT0000510 |
| Mouse | mmu-miR-320-3p | MIMAT0000666 |
| Rat | rno-miR-320-3p | MIMAT0000903 |
| Bovine | bta-miR-320a | MIMAT0003534 |
| Cricetulus griseus | cgr-miR-320a | MIMAT0023906 |
| Dog | cfa-miR-320 | MIMAT0006658 |
| View More Rows | ||
| Goat | chi-miR-320-3p | MIMAT0036132 |
| Gorilla gorilla | ggo-miR-320a | MIMAT0024080 |
| Microcebus murinus | mmr-miR-320a | MIMAT0049150 |
| Nomascus leucogenys | nle-miR-320a | MIMAT0049389 |
| Otolemur garnettii | oga-miR-320a | MIMAT0049611 |
| Pan paniscus | ppa-miR-320a | MIMAT0049065 |
| Pan troglodytes | ptr-miR-320a | MIMAT0008093 |
| Papio hamadryas | pha-miR-320a | MIMAT0049587 |
| Pongo pygmaeus | ppy-miR-320a | MIMAT0015821 |
| Pteropus alecto | pal-miR-320-3p | MIMAT0040074 |
| Rhesus monkey | mml-miR-320a | MIMAT0006263 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Dog
Goat
Gorilla gorilla
Microcebus murinus
Nomascus leucogenys
Otolemur garnettii
Pan paniscus
Pan troglodytes
Papio hamadryas
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-320a (v21)
Mouse: mmu-miR-320 (v17)
Rat: rno-miR-320 (v18)
Bos taurus: bta-miR-320 (v19)
Macaca mulatta: mml-miR-320 (v18)
Mouse: mmu-miR-320 (v17)
Rat: rno-miR-320 (v18)
Bos taurus: bta-miR-320 (v19)
Macaca mulatta: mml-miR-320 (v18)

Genomic Map