Assay Details
| Gene Family ID: | MIPF0000163, mir-320 | 
| Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCGA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-320a-3p | MIMAT0000510 | 
| Mouse | mmu-miR-320-3p | MIMAT0000666 | 
| Rat | rno-miR-320-3p | MIMAT0000903 | 
| Bovine | bta-miR-320a | MIMAT0003534 | 
| Cricetulus griseus | cgr-miR-320a | MIMAT0023906 | 
| Dog | cfa-miR-320 | MIMAT0006658 | 
| View More Rows | ||
| Goat | chi-miR-320-3p | MIMAT0036132 | 
| Gorilla gorilla | ggo-miR-320a | MIMAT0024080 | 
| Microcebus murinus | mmr-miR-320a | MIMAT0049150 | 
| Nomascus leucogenys | nle-miR-320a | MIMAT0049389 | 
| Otolemur garnettii | oga-miR-320a | MIMAT0049611 | 
| Pan paniscus | ppa-miR-320a | MIMAT0049065 | 
| Pan troglodytes | ptr-miR-320a | MIMAT0008093 | 
| Papio hamadryas | pha-miR-320a | MIMAT0049587 | 
| Pongo pygmaeus | ppy-miR-320a | MIMAT0015821 | 
| Pteropus alecto | pal-miR-320-3p | MIMAT0040074 | 
| Rhesus monkey | mml-miR-320a | MIMAT0006263 | 
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-320a | 
| Stem-loop Accession # | MI0000542 | 
| Stem-loop Sequence | CUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUG | 
| Chromosome Location | Chr. 8 - 22244966 - 22245037 [-] on Build GRCh38 | 
| Mature MicroRNA | miRBase Accession #: MIMAT0000510 miRBase ID: hsa-miR-320a-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: Chr. 8 - 22244966 - 22245037 [-] on Build GRCh38 | 
| Stem-loop ID | mmu-mir-320 | 
| Stem-loop Accession # | MI0000704 | 
| Stem-loop Sequence | GCCUCGCCGCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGUGGG | 
| Chromosome Location | Chr. 14 - 70443510 - 70443591 [+] on Build GRCm38 | 
| Mature MicroRNA | miRBase Accession #: MIMAT0000666 miRBase ID: mmu-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: Chr. 14 - 70443510 - 70443591 [+] on Build GRCm38 | 
| Stem-loop ID | rno-mir-320 | 
| Stem-loop Accession # | MI0000972 | 
| Stem-loop Sequence | GCCUCGCUGUCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUAUGGG | 
| Chromosome Location | Chr. 15 - 52134276 - 52134357 [+] on Build Rnor_6.0 | 
| Mature MicroRNA | miRBase Accession #: MIMAT0000903 miRBase ID: rno-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: Chr. 15 - 52134276 - 52134357 [+] on Build Rnor_6.0 | 
| Stem-loop ID | bta-mir-320a-1 | 
| Stem-loop Accession # | MI0012211 | 
| Stem-loop Sequence | CCCGCGGCGUCUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003534 miRBase ID: bta-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | bta-mir-320a-2 | 
| Stem-loop Accession # | MI0004748 | 
| Stem-loop Sequence | AAAAACGAAAAAGAGGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAAGAGGG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003534 miRBase ID: bta-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | cgr-mir-320a | 
| Stem-loop Accession # | MI0020491 | 
| Stem-loop Sequence | GCGCCUCGCUACCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAAAAAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0023906 miRBase ID: cgr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | cfa-mir-320 | 
| Stem-loop Accession # | MI0008063 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0006658 miRBase ID: cfa-miR-320 Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | chi-mir-320 | 
| Stem-loop Accession # | MI0030737 | 
| Stem-loop Sequence | GCGUCUCGCUCGCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAAGAGGGGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0036132 miRBase ID: chi-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | ggo-mir-320a | 
| Stem-loop Accession # | MI0020627 | 
| Stem-loop Sequence | GCCACAGUAUUUAUUAGGCGGCGCUUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGGUAACUGGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0024080 miRBase ID: ggo-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | mmr-mir-320a-1 | 
| Stem-loop Accession # | MI0039933 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049150 miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | mmr-mir-320a-2 | 
| Stem-loop Accession # | MI0039902 | 
| Stem-loop Sequence | GCAUUCUCUUCCCGGUUCUUCCCGGAGUCAGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049150 miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | mmr-mir-320a-3 | 
| Stem-loop Accession # | MI0039945 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049150 miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | mmr-mir-320a-4 | 
| Stem-loop Accession # | MI0039865 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049150 miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | nle-mir-320a | 
| Stem-loop Accession # | MI0040127 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049389 miRBase ID: nle-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | oga-mir-320a | 
| Stem-loop Accession # | MI0040392 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049611 miRBase ID: oga-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | ppa-mir-320a | 
| Stem-loop Accession # | MI0039773 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049065 miRBase ID: ppa-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | ptr-mir-320a | 
| Stem-loop Accession # | MI0008607 | 
| Stem-loop Sequence | GCUUCGCUCCUCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0008093 miRBase ID: ptr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | pha-mir-320a | 
| Stem-loop Accession # | MI0040365 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049587 miRBase ID: pha-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | ppy-mir-320a | 
| Stem-loop Accession # | MI0014884 | 
| Stem-loop Sequence | GCUUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGGU | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0015821 miRBase ID: ppy-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | pal-mir-320 | 
| Stem-loop Accession # | MI0032480 | 
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0040074 miRBase ID: pal-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
| Stem-loop ID | mml-mir-320a | 
| Stem-loop Accession # | MI0007691 | 
| Stem-loop Sequence | GCUUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0006263 miRBase ID: mml-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA | 
 Back To Top
 Back To TopMore Information
Related Products
 TaqMan™ Pri-miRNA Assay : Rn03465856_pri
						TaqMan™ Pri-miRNA Assay : Rn03465856_pri TaqMan™ Pri-miRNA Assay : Hs03303331_pri
						TaqMan™ Pri-miRNA Assay : Hs03303331_pri TaqMan™ Pri-miRNA Assay : Mm03307161_pri
						TaqMan™ Pri-miRNA Assay : Mm03307161_pri Ambion® Anti-miR™ miRNA Inhibitor : AM11621
						Ambion® Anti-miR™ miRNA Inhibitor : AM11621 Ambion® Pre-miR™ miRNA Precursor : PM11621
						Ambion® Pre-miR™ miRNA Precursor : PM11621 TaqMan™ MicroRNA Assay : 002277
						TaqMan™ MicroRNA Assay : 002277 mirVana® miRNA inhibitor : MH11621
						mirVana® miRNA inhibitor : MH11621 mirVana® miRNA mimic : MC11621
						mirVana® miRNA mimic : MC11621 TaqMan™ Advanced miRNA Assay : rno481048_mir
						TaqMan™ Advanced miRNA Assay : rno481048_mir TaqMan™ Advanced miRNA Assay : mmu481048_mir
						TaqMan™ Advanced miRNA Assay : mmu481048_mir FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
						FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2 MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Fast
						MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Fast FGMIR : TaqManTM Advanced miRNA Human A Card
						FGMIR : TaqManTM Advanced miRNA Human A Card MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
						MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
						MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast MIRNA : TaqMan® Advanced miRNA Human A Card
						MIRNA : TaqMan® Advanced miRNA Human A Card MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
						MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Standard
						MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls 96-well Plate, Standard MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
						MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
						FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
						MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast MIRNA : TaqMan® Advanced miRNA Human A and B Cards
						MIRNA : TaqMan® Advanced miRNA Human A and B Cards MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
						MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls Card
						FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls Card FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
						FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate
						FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
						FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2 FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
						FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2 MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
						MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
						FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2 FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate
						FGMIR : TaqManTM Advanced miRNA Human Endogenous Controls 96-well Plate MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls Card
						MIRNA : TaqMan® Advanced miRNA Human Endogenous Controls Card MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
						MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA PanelmiRBase Alias:
						
							Human: hsa-miR-320a (v21)
						
					
				 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top