Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-320a-3p | MIMAT0000510 |
| Mouse | mmu-miR-320-3p | MIMAT0000666 |
| Rat | rno-miR-320-3p | MIMAT0000903 |
| Bovine | bta-miR-320a | MIMAT0003534 |
| Cricetulus griseus | cgr-miR-320a | MIMAT0023906 |
| Dog | cfa-miR-320 | MIMAT0006658 |
| View More Rows | ||
| Goat | chi-miR-320-3p | MIMAT0036132 |
| Gorilla gorilla | ggo-miR-320a | MIMAT0024080 |
| Microcebus murinus | mmr-miR-320a | MIMAT0049150 |
| Nomascus leucogenys | nle-miR-320a | MIMAT0049389 |
| Otolemur garnettii | oga-miR-320a | MIMAT0049611 |
| Pan paniscus | ppa-miR-320a | MIMAT0049065 |
| Pan troglodytes | ptr-miR-320a | MIMAT0008093 |
| Papio hamadryas | pha-miR-320a | MIMAT0049587 |
| Pongo pygmaeus | ppy-miR-320a | MIMAT0015821 |
| Pteropus alecto | pal-miR-320-3p | MIMAT0040074 |
| Rhesus monkey | mml-miR-320a | MIMAT0006263 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-320a |
| Stem-loop Accession # | MI0000542 |
| Stem-loop Sequence | CUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUG |
| Chromosome Location | Chr. 8 - 22244966 - 22245037 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000510
miRBase ID: hsa-miR-320a-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: Chr. 8 - 22244966 - 22245037 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-320 |
| Stem-loop Accession # | MI0000704 |
| Stem-loop Sequence | GCCUCGCCGCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGUGGG |
| Chromosome Location | Chr. 14 - 70443510 - 70443591 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000666
miRBase ID: mmu-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: Chr. 14 - 70443510 - 70443591 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-320 |
| Stem-loop Accession # | MI0000972 |
| Stem-loop Sequence | GCCUCGCUGUCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUAUGGG |
| Chromosome Location | Chr. 15 - 52134276 - 52134357 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000903
miRBase ID: rno-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: Chr. 15 - 52134276 - 52134357 [+] on Build Rnor_6.0 |
| Stem-loop ID | bta-mir-320a-1 |
| Stem-loop Accession # | MI0012211 |
| Stem-loop Sequence | CCCGCGGCGUCUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003534
miRBase ID: bta-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | bta-mir-320a-2 |
| Stem-loop Accession # | MI0004748 |
| Stem-loop Sequence | AAAAACGAAAAAGAGGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAAGAGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003534
miRBase ID: bta-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | cgr-mir-320a |
| Stem-loop Accession # | MI0020491 |
| Stem-loop Sequence | GCGCCUCGCUACCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAAAAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023906
miRBase ID: cgr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | cfa-mir-320 |
| Stem-loop Accession # | MI0008063 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006658
miRBase ID: cfa-miR-320 Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | chi-mir-320 |
| Stem-loop Accession # | MI0030737 |
| Stem-loop Sequence | GCGUCUCGCUCGCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAAGAGGGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036132
miRBase ID: chi-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | ggo-mir-320a |
| Stem-loop Accession # | MI0020627 |
| Stem-loop Sequence | GCCACAGUAUUUAUUAGGCGGCGCUUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGGUAACUGGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0024080
miRBase ID: ggo-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | mmr-mir-320a-1 |
| Stem-loop Accession # | MI0039933 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049150
miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | mmr-mir-320a-2 |
| Stem-loop Accession # | MI0039902 |
| Stem-loop Sequence | GCAUUCUCUUCCCGGUUCUUCCCGGAGUCAGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049150
miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | mmr-mir-320a-3 |
| Stem-loop Accession # | MI0039945 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049150
miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | mmr-mir-320a-4 |
| Stem-loop Accession # | MI0039865 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049150
miRBase ID: mmr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | nle-mir-320a |
| Stem-loop Accession # | MI0040127 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049389
miRBase ID: nle-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | oga-mir-320a |
| Stem-loop Accession # | MI0040392 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049611
miRBase ID: oga-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | ppa-mir-320a |
| Stem-loop Accession # | MI0039773 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049065
miRBase ID: ppa-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-320a |
| Stem-loop Accession # | MI0008607 |
| Stem-loop Sequence | GCUUCGCUCCUCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008093
miRBase ID: ptr-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | pha-mir-320a |
| Stem-loop Accession # | MI0040365 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049587
miRBase ID: pha-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-320a |
| Stem-loop Accession # | MI0014884 |
| Stem-loop Sequence | GCUUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015821
miRBase ID: ppy-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | pal-mir-320 |
| Stem-loop Accession # | MI0032480 |
| Stem-loop Sequence | GCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0040074
miRBase ID: pal-miR-320-3p Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
| Stem-loop ID | mml-mir-320a |
| Stem-loop Accession # | MI0007691 |
| Stem-loop Sequence | GCUUCGCUCCCCUCCGCCUUCUCUUCCCGGUUCUUCCCGGAGUCGGGAAAAGCUGGGUUGAGAGGGCGAAAAAGGAUGAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006263
miRBase ID: mml-miR-320a Mature miRNA Sequence: AAAAGCUGGGUUGAGAGGGCGA Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Human: hsa-miR-320a (v21)

Genomic Map