Assay Details
Amplicon Length: | 78 |
Chromosome Location: | Chr. 19 - 53674003 - 53674003 [-] on 53674003 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-498-3p | MIMAT0037323 |
| Human | hsa-miR-498-5p | MIMAT0002824 |
Stem-loop Details
Human

Genomic Map