Assay Details
Gene Family ID: | MIPF0000463, mir-498 |
Mature miRNA Sequence: | UUUCAAGCCAGGGGGCGUUUUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-498-5p | MIMAT0002824 |
Pan troglodytes | ptr-miR-498 | MIMAT0008171 |
Pongo pygmaeus | ppy-miR-498 | MIMAT0015919 |
Rhesus monkey | mml-miR-498 | MIMAT0006360 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
