Assay Details
Amplicon Length: | 111 |
Chromosome Location: | Chr. 2 - 47377274 - 47377274 [+] on 47377274 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-559 | MIMAT0003223 |
Stem-loop Details
Human
