Assay Details
Gene Family ID: | MIPF0000656, mir-559 |
Mature miRNA Sequence: | UAAAGUAAAUAUGCACCAAAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-559 | MIMAT0003223 |
| Callithrix jacchus | cja-miR-559 | MIMAT0039614 |
| Pan troglodytes | ptr-miR-559 | MIMAT0008240 |
Stem-loop Details
Human
Callithrix jacchus
Pan troglodytes

Genomic Map