Assay Details
Amplicon Length: | 135 |
Chromosome Location: | Chr. 5 - 96078743 - 96078743 [-] on 96078743 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-583 | MIMAT0003248 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 135 |
Chromosome Location: | Chr. 5 - 96078743 - 96078743 [-] on 96078743 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-583 | MIMAT0003248 |