Assay Details
Gene Family ID: | MIPF0000482, mir-583 |
Mature miRNA Sequence: | CAAAGAGGAAGGUCCCAUUAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-583 | MIMAT0003248 |
| Pan troglodytes | ptr-miR-583 | MIMAT0008256 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map