Assay Details
| Amplicon Length: | 103 | 
| Chromosome Location: | Chr. 6 - 45197968 - 45197968 [-] on 45197968 Build GRCh38 | 
| Species: | Human | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-586 | MIMAT0003252 | 
Stem-loop Details
Human
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Ambion® Anti-miR™ miRNA Inhibitor :
						Ambion® Anti-miR™ miRNA Inhibitor :