Assay Details
Gene Family ID: | MIPF0000467, mir-586 |
Mature miRNA Sequence: | UAUGCAUUGUAUUUUUAGGUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-586 | MIMAT0003252 |
| Pongo pygmaeus | ppy-miR-586 | MIMAT0016028 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map