Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 8 - 104484866 - 104484866 [+] on 104484866 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548a-5p | MIMAT0004803 |
| Human | hsa-miR-548a-3p | MIMAT0003251 |
Stem-loop Details
Human

Genomic Map