Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAAUUGCGAGUUUUACC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548a-5p | MIMAT0004803 |
| Callithrix jacchus | cja-miR-548x | MIMAT0039577 |
Stem-loop Details
Human
Callithrix jacchus

Genomic Map