Assay Details
Amplicon Length: | 78 |
Chromosome Location: | Chr. 17 - 81124880 - 81124880 [+] on 81124880 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-657 | MIMAT0003335 |
Stem-loop Details
Human
