Assay Details
Gene Family ID: | MIPF0000498, mir-657 |
Mature miRNA Sequence: | GGCAGGUUCUCACCCUCUCUAGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-657 | MIMAT0003335 |
Pan troglodytes | ptr-miR-657 | MIMAT0008306 |
Stem-loop Details
Human
Pan troglodytes
