Assay Details
Gene Family ID: | MIPF0000185, mir-542 |
Mature miRNA Sequence: | UGUGACAGAUUGAUAACUGAAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-542-3p | MIMAT0003389 | |
| Mouse | mmu-miR-542-3p | MIMAT0003172 | |
| Rat | rno-miR-542-3p | MIMAT0003179 | |
| Dasypus novemcinctus | dno-miR-542-3p | MIMAT0047970 | |
| Dog | cfa-miR-542 | MIMAT0006747 | |
| Gorilla gorilla | ggo-miR-542 | MIMAT0024231 | |
| View More Rows | |||
| Guinea pig | cpo-miR-542-3p | MIMAT0047342 | |
| Horse | eca-miR-542-3p | MIMAT0013238 | |
| Microcebus murinus | mmr-miR-542 | MIMAT0049224 | |
| Nomascus leucogenys | nle-miR-542 | MIMAT0049392 | |
| Pig | ssc-miR-542-3p | MIMAT0013925 | |
| Pongo pygmaeus | ppy-miR-542-3p | MIMAT0015992 | |
| Pteropus alecto | pal-miR-542-3p | MIMAT0040184 | |
| Rabbit | ocu-miR-542-3p | MIMAT0048526 | |
| Rhesus monkey | mml-miR-542-3p | MIMAT0006407 | |
| Tupaia chinensis | tch-miR-542-3p | MIMAT0036613 | |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Horse
Microcebus murinus
Nomascus leucogenys
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Tupaia chinensis
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products

Genomic Map