Assay Details
Gene Family ID: | MIPF0000185, mir-542 |
Mature miRNA Sequence: | UGUGACAGAUUGAUAACUGAAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-542-3p | MIMAT0003389 |
| Mouse | mmu-miR-542-3p | MIMAT0003172 |
| Rat | rno-miR-542-3p | MIMAT0003179 |
| Dasypus novemcinctus | dno-miR-542-3p | MIMAT0047970 |
| Dog | cfa-miR-542 | MIMAT0006747 |
| Gorilla gorilla | ggo-miR-542 | MIMAT0024231 |
| View More Rows | ||
| Guinea pig | cpo-miR-542-3p | MIMAT0047342 |
| Horse | eca-miR-542-3p | MIMAT0013238 |
| Microcebus murinus | mmr-miR-542 | MIMAT0049224 |
| Nomascus leucogenys | nle-miR-542 | MIMAT0049392 |
| Pig | ssc-miR-542-3p | MIMAT0013925 |
| Pongo pygmaeus | ppy-miR-542-3p | MIMAT0015992 |
| Pteropus alecto | pal-miR-542-3p | MIMAT0040184 |
| Rabbit | ocu-miR-542-3p | MIMAT0048526 |
| Rhesus monkey | mml-miR-542-3p | MIMAT0006407 |
| Tupaia chinensis | tch-miR-542-3p | MIMAT0036613 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-542 |
| Stem-loop Accession # | MI0003686 |
| Stem-loop Sequence | CAGAUCUCAGACAUCUCGGGGAUCAUCAUGUCACGAGAUACCAGUGUGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGAGCCACUCAUCUUCA |
| Chromosome Location | Chr. X - 134541341 - 134541437 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003389
miRBase ID: hsa-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: Chr. X - 134541341 - 134541437 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-542 |
| Stem-loop Accession # | MI0003522 |
| Stem-loop Sequence | GGAUCUCAGACGUCUCGGGGAUCAUCAUGUCACGAGAUACCACUGUGCCCUUGUGACAGAUUGAUAACUGAAAGGUCUGGGAGCC |
| Chromosome Location | Chr. X - 53049403 - 53049487 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003172
miRBase ID: mmu-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: Chr. X - 53049403 - 53049487 [-] on Build GRCm38 |
| Stem-loop ID | rno-mir-542-1 |
| Stem-loop Accession # | MI0003528 |
| Stem-loop Sequence | UCUCAGACGUCUCGGGGAUCAUCAUGUCACGAGAUACCACUGCGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGA |
| Chromosome Location | Chr. X - 158152146 - 158152224 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003179
miRBase ID: rno-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: Chr. X - 158588529 - 158588607 [+] on Build Rnor_6.0 |
| Stem-loop ID | rno-mir-542-2 |
| Stem-loop Accession # | MI0031781 |
| Stem-loop Sequence | UCUCAGACGUCUCGGGGAUCAUCAUGUCACGAGAUACCACUGCGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGA |
| Chromosome Location | Chr. X - 158158885 - 158158963 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003179
miRBase ID: rno-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: Chr. X - 158588529 - 158588607 [+] on Build Rnor_6.0 |
| Stem-loop ID | rno-mir-542-3 |
| Stem-loop Accession # | MI0031782 |
| Stem-loop Sequence | UCUCAGACGUCUCGGGGAUCAUCAUGUCACGAGAUACCACUGCGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGA |
| Chromosome Location | Chr. X - 158588529 - 158588607 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003179
miRBase ID: rno-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: Chr. X - 158588529 - 158588607 [+] on Build Rnor_6.0 |
| Stem-loop ID | dno-mir-542 |
| Stem-loop Accession # | MI0039151 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACGAGAUACCCACUGUGCACUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047970
miRBase ID: dno-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | cfa-mir-542 |
| Stem-loop Accession # | MI0008171 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACGAGAUACCACUGUGCACUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006747
miRBase ID: cfa-miR-542 Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | ggo-mir-542 |
| Stem-loop Accession # | MI0020778 |
| Stem-loop Sequence | GGAUCGGUCAAGGAUGCACAGAUCUCAGACAUCUCGGGGAUCAUCAUGUCACGAGAUACCAGUGUGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGAGCCACUCAUCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0024231
miRBase ID: ggo-miR-542 Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | cpo-mir-542 |
| Stem-loop Accession # | MI0038820 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACCAGAUACCAUUGUGCGCUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047342
miRBase ID: cpo-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | eca-mir-542 |
| Stem-loop Accession # | MI0012980 |
| Stem-loop Sequence | AGGACCUCAGACAUCUCGGGGAUCAUCAUGUCACGAGAUACCACUGUGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGAGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013238
miRBase ID: eca-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | mmr-mir-542 |
| Stem-loop Accession # | MI0039951 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACGAGAUACCACUGUGCACUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049224
miRBase ID: mmr-miR-542 Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | nle-mir-542 |
| Stem-loop Accession # | MI0040130 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACGAGAUACCAGUGUGCACUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0049392
miRBase ID: nle-miR-542 Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | ssc-mir-542 |
| Stem-loop Accession # | MI0013138 |
| Stem-loop Sequence | CCUCAGACGUCUCGGGGAUCAUCAUGUCACGAGAGACCACUGUGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013925
miRBase ID: ssc-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-542 |
| Stem-loop Accession # | MI0015040 |
| Stem-loop Sequence | CAGAUCUCAGACAUCUCGGGGAUCAUCAUGUCACGAGAUACCAGUGUGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGAGCCACUCAUCUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015992
miRBase ID: ppy-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | pal-mir-542 |
| Stem-loop Accession # | MI0032538 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACGAGAGACCAUUGUGCACUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0040184
miRBase ID: pal-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | ocu-mir-542 |
| Stem-loop Accession # | MI0039445 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACGAGAUACCACUGUGCACUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048526
miRBase ID: ocu-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | mml-mir-542 |
| Stem-loop Accession # | MI0007816 |
| Stem-loop Sequence | CAGACCUCAGACAUCUCGGGGAUCAUCAUGUCACGAGAUACCACUGUGCACUUGUGACAGAUUGAUAACUGAAAGGUCUGGGAGCCAUUCAUCUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006407
miRBase ID: mml-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
| Stem-loop ID | tch-mir-542 |
| Stem-loop Accession # | MI0031283 |
| Stem-loop Sequence | UCGGGGAUCAUCAUGUCACGAGAUACCACUGCGCGCUUGUGACAGAUUGAUAACUGAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036613
miRBase ID: tch-miR-542-3p Mature miRNA Sequence: UGUGACAGAUUGAUAACUGAAA Chromosome Location: NA |
More Information
Related Products

Genomic Map