Assay Details
Gene Family ID: | MIPF0000185, mir-542 |
Mature miRNA Sequence: | UGUGACAGAUUGAUAACUGAAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-542-3p | MIMAT0003389 |
| Mouse | mmu-miR-542-3p | MIMAT0003172 |
| Rat | rno-miR-542-3p | MIMAT0003179 |
| Dasypus novemcinctus | dno-miR-542-3p | MIMAT0047970 |
| Dog | cfa-miR-542 | MIMAT0006747 |
| Gorilla gorilla | ggo-miR-542 | MIMAT0024231 |
| View More Rows | ||
| Guinea pig | cpo-miR-542-3p | MIMAT0047342 |
| Horse | eca-miR-542-3p | MIMAT0013238 |
| Microcebus murinus | mmr-miR-542 | MIMAT0049224 |
| Nomascus leucogenys | nle-miR-542 | MIMAT0049392 |
| Pig | ssc-miR-542-3p | MIMAT0013925 |
| Pongo pygmaeus | ppy-miR-542-3p | MIMAT0015992 |
| Pteropus alecto | pal-miR-542-3p | MIMAT0040184 |
| Rabbit | ocu-miR-542-3p | MIMAT0048526 |
| Rhesus monkey | mml-miR-542-3p | MIMAT0006407 |
| Tupaia chinensis | tch-miR-542-3p | MIMAT0036613 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Horse
Microcebus murinus
Nomascus leucogenys
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Tupaia chinensis

Genomic Map