Assay Details
Amplicon Length: | 57 |
Chromosome Location: | Chr. 1 - 154194066 - 154194066 [+] on 154194066 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-190b-5p | MIMAT0004929 |
| Human | hsa-miR-190b-3p | MIMAT0037332 |
Stem-loop Details
Human

Genomic Map