Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | UGAUAUGUUUGAUAUUGGGUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-190b-5p | MIMAT0004929 |
| Mouse | mmu-miR-190b-5p | MIMAT0004852 |
| Callithrix jacchus | cja-miR-190b | MIMAT0039487 |
| Cricetulus griseus | cgr-miR-190b | MIMAT0023821 |
| Dasypus novemcinctus | dno-miR-190b-5p | MIMAT0047715 |
| Goat | chi-miR-190b | MIMAT0036021 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-190b | MIMAT0024270 |
| Guinea pig | cpo-miR-190b-5p | MIMAT0047064 |
| Pig | ssc-miR-190b | MIMAT0020588 |
| Rabbit | ocu-miR-190b-5p | MIMAT0048301 |
Stem-loop Details
Human
Mouse
Callithrix jacchus
Cricetulus griseus
Dasypus novemcinctus
Goat
Gorilla gorilla
Guinea pig
Pig
Rabbit
More Information
Related Products
miRBase Alias:
Staphylococcus schleiferi: ssc-miR-190 (v18)

Genomic Map