Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 10 - 12725105 - 12725105 [+] on 12725105 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548q | MIMAT0011163 |
Stem-loop Details
Human
