Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | GCUGGUGCAAAAGUAAUGGCGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548q | MIMAT0011163 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-548q |
---|---|
miRBase Accession Number: | MI0010637 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GCUGGUGCAAAAGUAAUGGCGG |
Species: | Human |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | GCUGGUGCAAAAGUAAUGGCGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548q | MIMAT0011163 |