Assay Details
Mature miRNA Sequence: | GGCAUGAUGUAGCAGUGGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-270 | MIMAT0000326 |
Stem-loop Details
Caenorhabditis elegans
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cel-miR-270 |
|---|---|
| miRBase Accession Number: | MI0000350 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGCAUGAUGUAGCAGUGGAG |
| Species: | Caenorhabditis elegans |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | GGCAUGAUGUAGCAGUGGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-270 | MIMAT0000326 |