Assay Details
Mature miRNA Sequence: | GGCAUGAUGUAGCAGUGGAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis elegans | cel-miR-270 | MIMAT0000326 |
Stem-loop Details
Caenorhabditis elegans

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | cel-miR-270 |
---|---|
miRBase Accession Number: | MI0000350 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GGCAUGAUGUAGCAGUGGAG |
Species: | Caenorhabditis elegans |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | GGCAUGAUGUAGCAGUGGAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis elegans | cel-miR-270 | MIMAT0000326 |