Assay Details
Gene Family ID: | MIPF0000470, mir-549 |
Mature miRNA Sequence: | UGACAACUAUGGAUGAGCUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-549a-3p | MIMAT0003333 |
| Pan troglodytes | ptr-miR-549 | MIMAT0008229 |
| Pongo pygmaeus | ppy-miR-549 | MIMAT0015996 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
More Information
Related Products
miRBase Alias:
Human: hsa-miR-549a (v21)
Human: hsa-miR-549 (v18)
Human: hsa-miR-549 (v18)

Genomic Map