Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 15 - 80841776 - 80841776 [+] on 80841776 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-549a-5p | MIMAT0037328 |
| Human | hsa-miR-549a-3p | MIMAT0003333 |
Stem-loop Details
Human

Genomic Map