Assay Details
Gene Family ID: | MIPF0000162, mir-367 |
Mature miRNA Sequence: | ACUGUUGCUAAUAUGCAACUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-367-5p | MIMAT0004686 |
| Callithrix jacchus | cja-miR-367 | MIMAT0039585 |
| Dasypus novemcinctus | dno-miR-367-5p | MIMAT0047838 |
| Dog | cfa-miR-367 | MIMAT0009859 |
| Rabbit | ocu-miR-367-5p | MIMAT0036324 |
Stem-loop Details
Human
Callithrix jacchus
Dasypus novemcinctus
Dog
Rabbit

Genomic Map