Assay Details
Amplicon Length: | 131 |
Chromosome Location: | Chr. 4 - 112647628 - 112647628 [-] on 112647628 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302d-5p | MIMAT0004685 |
| Human | hsa-miR-302d-3p | MIMAT0000718 |
| Human | hsa-miR-367-5p | MIMAT0004686 |
| Human | hsa-miR-367-3p | MIMAT0000719 |
Stem-loop Details
Human

Genomic Map