Assay Details
| Gene Family ID: | MIPF0000386, mir-743 | 
| Mature miRNA Sequence: | GAAAGACAUCAUGCUGAAUAGA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-743b-3p | MIMAT0004840 | 
Stem-loop Details
Mouse
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-743b-3p | 
|---|---|
| miRBase Accession Number: | MI0005470 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | GAAAGACAUCAUGCUGAAUAGA | 
| Species: | Mouse | 
| Product Type: | mirVana® miRNA mimic | 
| Gene Family ID: | MIPF0000386, mir-743 | 
| Mature miRNA Sequence: | GAAAGACAUCAUGCUGAAUAGA | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-743b-3p | MIMAT0004840 | 
 Back To Top
 Back To Top