Assay Details
Amplicon Length: | 85 |
Chromosome Location: | Chr. X - 66777755 - 66777755 [+] on 66777755 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-743b-3p | MIMAT0004840 |
| Mouse | mmu-miR-743b-5p | MIMAT0004839 |
Stem-loop Details
Mouse

Genomic Map