Assay Details
Gene Family ID: | MIPF0001000, MIR2109 |
Mature miRNA Sequence: | UGCGAGUGUCUUCGCCUCUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Glycine max | gma-miR2109-5p | MIMAT0010086 |
Stem-loop Details
Glycine max
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | gma-miR2109-5p |
|---|---|
| miRBase Accession Number: | MI0010579 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGCGAGUGUCUUCGCCUCUG |
| Species: | Glycine max |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0001000, MIR2109 |
Mature miRNA Sequence: | UGCGAGUGUCUUCGCCUCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Glycine max | gma-miR2109-5p | MIMAT0010086 |