Assay Details
Gene Family ID: | MIPF0001000, MIR2109 |
Mature miRNA Sequence: | UGCGAGUGUCUUCGCCUCUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Glycine max | gma-miR2109-5p | MIMAT0010086 |
Stem-loop Details
Glycine max

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | gma-miR2109-5p |
---|---|
miRBase Accession Number: | MI0010579 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGCGAGUGUCUUCGCCUCUG |
Species: | Glycine max |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001000, MIR2109 |
Mature miRNA Sequence: | UGCGAGUGUCUUCGCCUCUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Glycine max | gma-miR2109-5p | MIMAT0010086 |