Assay Details
| Mature miRNA Sequence: | UCCCGUGGAGGCAGCCGAUG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Rice | osa-miR2098-5p | MIMAT0010060 | 
Stem-loop Details
Rice
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | osa-miR2098-5p | 
|---|---|
| miRBase Accession Number: | MI0010558 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | UCCCGUGGAGGCAGCCGAUG | 
| Species: | Rice | 
| Product Type: | mirVana® miRNA mimic | 
| Mature miRNA Sequence: | UCCCGUGGAGGCAGCCGAUG | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Rice | osa-miR2098-5p | MIMAT0010060 | 
 Back To Top
 Back To Top