Assay Details
Mature miRNA Sequence: | UCCCGUGGAGGCAGCCGAUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rice | osa-miR2098-5p | MIMAT0010060 |
Stem-loop Details
Rice
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | osa-miR2098-5p |
|---|---|
| miRBase Accession Number: | MI0010558 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCGUGGAGGCAGCCGAUG |
| Species: | Rice |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UCCCGUGGAGGCAGCCGAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rice | osa-miR2098-5p | MIMAT0010060 |