Assay Details
Mature miRNA Sequence: | UUUGUUACUGAUGGUGCUGUC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2575-5p | MIMAT0012420 | 
Stem-loop Details
Drosophila pseudoobscura
More Information
Related Products
miRBase Alias:
						
							Derxomyces pseudoschimicola: dps-miR-2575 (v18)
						
					
				
Genomic Map