Assay Details
Mature miRNA Sequence: | UUUGUUACUGAUGGUGCUGUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila pseudoobscura | dps-miR-2575-5p | MIMAT0012420 |
Stem-loop Details
Drosophila pseudoobscura

More Information
Related Products
miRBase Alias:
Derxomyces pseudoschimicola: dps-miR-2575 (v18)
