Assay Details
Mature miRNA Sequence: | CGUCUUACACUGCUGUGCUAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Hydra magnipapillata | hma-miR-2030 | MIMAT0014684 |
Stem-loop Details
Hydra magnipapillata
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hma-miR-2030 |
|---|---|
| miRBase Accession Number: | MI0013877 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGUCUUACACUGCUGUGCUAC |
| Species: | Hydra magnipapillata |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CGUCUUACACUGCUGUGCUAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Hydra magnipapillata | hma-miR-2030 | MIMAT0014684 |