Assay Details
Mature miRNA Sequence: | CGUCUUACACUGCUGUGCUAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Hydra magnipapillata | hma-miR-2030 | MIMAT0014684 |
Stem-loop Details
Hydra magnipapillata

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hma-miR-2030 |
---|---|
miRBase Accession Number: | MI0013877 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CGUCUUACACUGCUGUGCUAC |
Species: | Hydra magnipapillata |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | CGUCUUACACUGCUGUGCUAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Hydra magnipapillata | hma-miR-2030 | MIMAT0014684 |