Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUUGCCCAUGGUGUGUUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-496a-5p | MIMAT0017244 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-496-5p (v18)
Mouse: mmu-miR-496* (v17)
Mouse: mmu-miR-496* (v17)

Genomic Map