Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. 12 - 109739518 - 109739518 [+] on 109739518 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-496a-5p | MIMAT0017244 |
Mouse | mmu-miR-496a-3p | MIMAT0003738 |
Stem-loop Details
Mouse
