Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-499a | MIMAT0024118 |
Stem-loop Details
Gorilla gorilla
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ggo-miR-499a |
|---|---|
| miRBase Accession Number: | MI0020665 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUU |
| Species: | Gorilla gorilla |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-499a | MIMAT0024118 |