Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Gorilla gorilla | ggo-miR-499a | MIMAT0024118 |
Stem-loop Details
Gorilla gorilla

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ggo-miR-499a |
---|---|
miRBase Accession Number: | MI0020665 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUU |
Species: | Gorilla gorilla |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Gorilla gorilla | ggo-miR-499a | MIMAT0024118 |