Assay Details
Gene Family ID: | MIPF0001437, mir-2763 |
Mature miRNA Sequence: | UAUUAUGCUCAUUACUUUGGAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Heliconius melpomene | hme-miR-2763 | MIMAT0024955 |
Stem-loop Details
Heliconius melpomene
