Assay Details
Gene Family ID: | MIPF0001437, mir-2763 |
Mature miRNA Sequence: | UAUUAUGCUCAUUACUUUGGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Heliconius melpomene | hme-miR-2763 | MIMAT0024955 |
Stem-loop Details
Heliconius melpomene
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hme-miR-2763 |
|---|---|
| miRBase Accession Number: | MI0021750 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUUAUGCUCAUUACUUUGGAG |
| Species: | Heliconius melpomene |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001437, mir-2763 |
Mature miRNA Sequence: | UAUUAUGCUCAUUACUUUGGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Heliconius melpomene | hme-miR-2763 | MIMAT0024955 |