Assay Details
Mature miRNA Sequence: | UCAGUUAUCACAGUGCUGAUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-101-5p | MIMAT0038116 |
| Chicken | gga-miR-101-2-5p | MIMAT0031077 |
| Chrysemys picta | cpi-miR-101-5p | MIMAT0037716 |
| Columba livia | cli-miR-101-5p | MIMAT0038482 |
| Dasypus novemcinctus | dno-miR-101-5p | MIMAT0047598 |
| Gadus morhua | gmo-miR-101a-5p | MIMAT0044119 |
| View More Rows | ||
| Guinea pig | cpo-miR-101-5p | MIMAT0046947 |
| Monodelphis domestica | mdo-miR-101-1-5p | MIMAT0031076 |
| Rabbit | ocu-miR-101-5p | MIMAT0048186 |
| Rhesus monkey | mml-miR-101-1-5p | MIMAT0026581 |
Stem-loop Details
Alligator mississippiensis
Chicken
Chrysemys picta
Columba livia
Dasypus novemcinctus
Gadus morhua
Guinea pig
Monodelphis domestica
Rabbit
Rhesus monkey

Genomic Map