Assay Details
Mature miRNA Sequence: | UCAGUUAUCACAGUGCUGAUGCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Alligator mississippiensis | ami-miR-101-5p | MIMAT0038116 |
Chicken | gga-miR-101-2-5p | MIMAT0031077 |
Chrysemys picta | cpi-miR-101-5p | MIMAT0037716 |
Columba livia | cli-miR-101-5p | MIMAT0038482 |
Dasypus novemcinctus | dno-miR-101-5p | MIMAT0047598 |
Gadus morhua | gmo-miR-101a-5p | MIMAT0044119 |
View More Rows | ||
Guinea pig | cpo-miR-101-5p | MIMAT0046947 |
Monodelphis domestica | mdo-miR-101-1-5p | MIMAT0031076 |
Rabbit | ocu-miR-101-5p | MIMAT0048186 |
Rhesus monkey | mml-miR-101-1-5p | MIMAT0026581 |
Stem-loop Details
Alligator mississippiensis
Chicken
Chrysemys picta
Columba livia
Dasypus novemcinctus
Gadus morhua
Guinea pig
Monodelphis domestica
Rabbit
Rhesus monkey
