Assay Details
Gene Family ID: | MIPF0000040, mir-199 |
Mature miRNA Sequence: | CCCAGUGUUUAGACUACCUGUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-199b-5p | MIMAT0000672 |
Cricetulus griseus | cgr-miR-199b | MIMAT0023838 |
Stem-loop Details
Mouse
Cricetulus griseus
