Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 2 - 32318963 - 32318963 [+] on 32318963 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-199b-5p | MIMAT0000672 |
Mouse | mmu-miR-199b-3p | MIMAT0004667 |
Mouse | mmu-miR-3154 | MIMAT0035714 |
Stem-loop Details
Mouse
